Pfefferminzöl

Lieferant: CAESAR LORENTZ

G474.0025
CSLOG474.0025EA 28.6 EUR
CSLOG474.0025
Pfefferminzöl
Peppermint oil
MDL: MFCD00147870
CAS-Nummer: 8006-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR