N-Desmethylcitalopram-D3 hydrochloride solution, certified reference material, Supelco®
Lieferant: Merck
D-074-1ML
D-074-1MLEA 234 EUR
D-074-1ML
N-Desmethylcitalopram-D3 hydrochloride solution, certified reference material, Supelco®
Standards Organische Standards
A stable-labeled internal standard of N-Desmethylcitalopram, an active metabolite of the SSRI antidepressant drug citalopram.
Verpackung: Snap-N-Spike®/Snap-N-Shoot®
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=359ecdcb4729cd974fab1943292757c05b7bf8c0&vid=b566759bc2e651abd2554958832ba021a84e6723&prid=web&clid=SAPDAM