N-Desmethylcitalopram-D3 hydrochloride solution, certified reference material, Supelco®
Lieferant: Merck
D-074-1ML
D-074-1MLEA
234
EUR
D-074-1ML
N-Desmethylcitalopram-D3 hydrochloride solution, certified reference material, Supelco®
Standards
Organische Standards
A stable-labeled internal standard of N-Desmethylcitalopram, an active metabolite of the SSRI antidepressant drug citalopram.
Verpackung: Snap-N-Spike®/Snap-N-Shoot®