Salzsäure Konzentrat zur Verdünnung zu 0.01 mol (0.01N), DILUT-IT® Auflösungsmedium-Konzentrat, J.T.Baker®

Lieferant: Avantor
Gefahr

D010-00EA 31.98 EUR
D010-00 D010-02 D010-04
Salzsäure Konzentrat zur Verdünnung zu 0.01 mol (0.01N), DILUT-IT® Auflösungsmedium-Konzentrat, J.T.Baker®
Salzsäure

DILUT-IT dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities

Formel: HCl
Dichte: 1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use Passes Test
STORE AT ROOM TEMPERATURE For Information Only
Appearance Passes Test
Density (g/mL) at 25°C, For Information Only
Normality at 26x dilution 0.0098 - 0.0102
DILUT-IT dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR