Salzsäure Konzentrat zur Verdünnung zu 0.1 mol (0.1N), DILUT-IT® Auflösungsmedium-Konzentrat, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Hydrochloric acid solution, Muriatic acid

D011-00EA 32.1 EUR
D011-00 D011-02 D011-04
Salzsäure Konzentrat zur Verdünnung zu 0.1 mol (0.1N), DILUT-IT® Auflösungsmedium-Konzentrat, J.T.Baker®
Salzsäure

DILUT-IT dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities

Formel: HCl
Molecular Weight: 36.46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use 0.098 - 0.102
STORE AT ROOM TEMPERATURE
Appearance Passes Test
Density (g/mL) at 25°C, For Information Only
Normality at 26x dilution 0.098 - 0.102
DILUT-IT dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR