Kaliumdihydrogenphosphat Konzentrat zur Verdünnung zu pH 7,4, DILUT-IT® Auflösungsmedium-Konzentrat, J.T.Baker®

Lieferant: Avantor

D019-00EA 0 EUR
D019-00 D019-04
Kaliumdihydrogenphosphat Konzentrat zur Verdünnung zu pH 7,4, DILUT-IT® Auflösungsmedium-Konzentrat, J.T.Baker®
Kaliumdihydrogenphosphat

DILUT-IT dissolution media concentrates are produced from raw materials in accordance with USP/Ph. Eur./BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities.

Formel: KH₂PO₄
Dichte: 1,0000...2.0000 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011401
CAS-Nummer: 7778-77-0

Bestellen

Spezifikation Testergebnisse

Assay (M) at 26X Dilution (as Potassium Phosphate Monobasic) 0.045 - 0.054
Appearance Passes Test
pH at 25 °C at 26X Dilution 7.35 - 7.45
For Laboratory, Research or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR