5-DTAF hydrochloride, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 5-[(4,6-Dichlorotriazin-2-yl)amino]fluorescein hydrochloride

D0531-100MG D0531-1G D0531-500MG
D0531-100MGEA 131 EUR
D0531-100MG D0531-1G D0531-500MG
5-DTAF hydrochloride, Sigma-Aldrich®
5-DTAF hydrochloride

5-([4,6-Dichlorotriazin-2-yl]amino)fluorescein hydrochloride has been used for labeling of yeast cells to study the adhesion of yeast cells to the digestive tract of sea bass (Dicentrarchus labrax) larvae and leopard grouper (Mycteroperca rosacea).

Formel: C₂₃H₁₂Cl₂N₄O₅·HCl
Molecular Weight: 531.8 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00051407
CAS-Nummer: 21811-74-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8811c83714813ba040ff1986a2739d1d80f994eb&vid=373ab5126d99764233de3d92383391a92c959272&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR