Dekalzifizierende Lösung-Lite in wässriger Lösung

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

D0818-1L
D0818-1LEA 56.3 EUR
D0818-1L
Dekalzifizierende Lösung-Lite in wässriger Lösung
Decalcifying Solution-Lite

Hydrochloric acid based aqueous solution. Decalcifying Solution-Lite is designed to be a universal effective decalcifying agent. It is intended to be used for the decalcification of routine, immunohistochemical and bone marrow core specimens.


Decalcifying Solution-Lite has been tailored to suit your specific lab routine. It works equally well with all types of specimens in an easy to handle time frame.

Siedepunkt: 208 °F
Lagertemperatur: Raumtemperatur
UN: 1789
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to amber water like liquid with suspended dark particulate that will accumulate and intesify over time.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR