p-Xylene-bis(N-pyridinium bromide) (DPX) ≥95% (durch HPLC), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
D0940-500MG
D0940-500MGEA
229
EUR
D0940-500MG
p-Xylene-bis(N-pyridinium bromide) (DPX) ≥95% (durch HPLC), Sigma-Aldrich®
p-Xylene-bis(N-pyridinium bromide) (DPX)
p-Xylene-bis(N-pyridinium bromide). ≥95% (TLC) (DPX)
Formel:
C₁₈H₁₈Br₂N₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD00043749 CAS-Nummer: 14208-10-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...