Di-8-ANEPPS ≥95% (durch HPLC), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

D1065-5MG
D1065-5MGEA 733 EUR
D1065-5MG
Di-8-ANEPPS ≥95% (durch HPLC), Sigma-Aldrich®
Di-8-ANEPPS

4-(2-[6-(Dioctylamino)-2-naphthalenyl]ethenyl)-1-(3-sulfopropyl)pyridinium inner salt. ≥95% (HPLC), solid (Di-8-ANEPPS)

Formel: C₃₆H₅₂N₂O₃S
Lagertemperatur: Kühlschrank
MDL: MFCD03453608
CAS-Nummer: 157134-53-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd682125a1455f5352a6a6e0f70ff833e3bbbd7b&vid=643a425ea4e15588beb6a1e428b4c136eaa72a6f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR