Density gradient medium, OptiPrep™
Lieferant: SIGMA ALDRICH MICROSCOPY
D1556-250ML
D1556-250MLEA 606 EUR
D1556-250ML
Density gradient medium, OptiPrep™
Cell Separation Media
Density gradient medium is suitable for cell and subcellular organelle isolation, virus purification, and isolation of lipoproteins and macromolecules.
OptiPrep™ Density Gradient Medium has been used in the non enveloped virion isolation from bronchoalveolar lavage.