3,5-Dinitroanilin 97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

D193402-10G
D193402-10GEA 305 EUR
D193402-10G
3,5-Dinitroanilin 97%, Sigma-Aldrich®
3,5-Dinitroanilin
Formel: C₆H₅N₃O₄
Dichte: 1,601 g/cm³ (20 °C)
Lagertemperatur: Room Temperature
MDL: MFCD00007263
CAS-Nummer: 618-87-1
EINECS: 210-567-8
UN: 1596
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=366a70891a50dfc3fce5f0297dd44dcf93962d9f&vid=09803a9396a8b489a22c42e41d5e7a5fb0732f6f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Faint Yellow to Dark Yellow and Faint Brown to Dark Brown
Appearance (Form) Powder or Crystals or Powder with Chunk(s) or Crystals with Chunk(s)
Infrared spectrum Conforms to Structure
Carbon 38.0 - 40.7 %
Nitrogen 22.1 - 23.7 %
Purity (GC) ≥96.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR