Histodenz™, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Iohexol, N1,N3-Bis(2,3-dihydroxypropyl)-5-(N-(2,3-dihydroxypropyl)acetamido)-2,4,6-triiodoisophthalamide, 5-[Acetyl(2,3-dihydroxypropyl)amino]-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide, N,N'-Bis(2,3-dihydroxypropyl)-5-[N-(2,3-dihydroxypropyl)acetamido]-2,4,6-triiodoisophthalamide

D2158-100G
D2158-100GEA 516 EUR
D2158-100G
Histodenz™, Sigma-Aldrich®
5-(N-2-3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide

Useful as a nonionic density gradient medium.

Formel: C₁₉H₂₆I₃N₃O₉
Molecular Weight: 821,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077732
CAS-Nummer: 66108-95-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc4f1dfd3cdb79120e6cb496a5f5aeea4443f221&vid=8bd84de3f3b311dddcbc3883afc702a9e47ff749&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR