DAF-2 diacetate, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 2-​(3,​6-​diacetyloxy-​4,​5-​diamino-​9H-​xanthen-​9-​yl)-​benzoic acid, DAF-2 DA, 4,5-Diaminofluorescein diacetate

D2813-1MG
D2813-1MGEA 728 EUR
D2813-1MG
DAF-2 diacetate, Sigma-Aldrich®
DAF-2 diacetate

Cell permeable fluorescent detector of nitric oxide in living cells. 4,5-Diaminofluorescein diacetate (DAF-2 DA) has been used as a fluorescent dye for NO (nitric oxide) detection.

Formel: C₂₄H₁₈N₂O₇
Lagertemperatur: Kühlschrank
MDL: MFCD05865101
CAS-Nummer: 205391-02-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5b77754c28fac9bce6ab45ea04ed657b16ca006&vid=52c3eaddbc4022e2a46ee16a9392b839f5713713&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR