3,3'-Dioctadecyloxacarbocyanine perchlorate (DiO), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
DiO perchlorate, 3,3-Dioctadecyloxacarbocyanine perchlorate
D4292-20MG
D4292-20MGEA
180
EUR
D4292-20MG
3,3'-Dioctadecyloxacarbocyanine perchlorate (DiO), Sigma-Aldrich®
3,3'-Dioctadecyloxacarbocyanine perchlorate (DiO)
3,3′-Dioctadecyloxacarbocyanine perchlorate has been used for lipid bilayer labeling in antigen-presenting cells. It has also been used as a lipophilic dye for labeling in zebrafish embryos.
Formel:
C₅₃H₈₅ClN₂O₆ Lagertemperatur: Kühlschrank |
CAS-Nummer:
34215-57-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...