4-(Dimethylamino)zimtaldehyd chromogenes Reagenz für Indole und Flavanole, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
Reagent for assay of indole product of apotryptophanase and tryptophanase. DMAC produces colored adducts with flavanols for subsequent HPLC. It has been used in a chromogenic method for quantifying proanthocyandines in cranberry powder.
- 4-(Dimethylamino)cinnamaldehyde (DMAC) has been used in the serine protease inhibition assay
- It has been used for the detection of (epi)-flavan-3-ols and proanthocyanidin polymers in Brassica napus seeds
- It has been used for the determination of phenolic compounds in leaf samples
- It has been used for the staining of flavan-3-ols in plant samples
Formel:
C₁₁H₁₃NO Schmelzpunkt: 138,..140 °C (Lit.) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00007002 CAS-Nummer: 6203-18-5 EINECS: 228-267-0 |
|
Spezifikation Testergebnisse
Appearance (colour) | Yellow to very dark yellow and orange to very dark orange |
Appearance (form) | Powder |
Purity (HPLC area %) | ≥97.5 % |
Solubility (colour) | Yellow to very dark yellow to very dark red |
Solubility (turbidity) | Clear to hazy |
Solubility (method) | 250 mg in 5 ml chloroform/ethanol (1:1) |
Proton nmr spectrum | Conforms to structure |
Recommended retest period | 48 months |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...