4-(Dimethylamino)zimtaldehyd chromogenes Reagenz für Indole und Flavanole, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung


M-Clarity™ Qualitätsstufe = MQ200

D4506-10G D4506-1G D4506-25G D4506-5G
D4506-10GEA 210 EUR
D4506-10G D4506-1G D4506-25G D4506-5G
4-(Dimethylamino)zimtaldehyd chromogenes Reagenz für Indole und Flavanole, Sigma-Aldrich®
4-(Dimethylamino)zimtaldehyd

Reagent for assay of indole product of apotryptophanase and tryptophanase. DMAC produces colored adducts with flavanols for subsequent HPLC. It has been used in a chromogenic method for quantifying proanthocyandines in cranberry powder.


  • 4-(Dimethylamino)cinnamaldehyde (DMAC) has been used in the serine protease inhibition assay
  • It has been used for the detection of (epi)-flavan-3-ols and proanthocyanidin polymers in Brassica napus seeds
  • It has been used for the determination of phenolic compounds in leaf samples
  • It has been used for the staining of flavan-3-ols in plant samples

Formel: C₁₁H₁₃NO
Schmelzpunkt: 138,..140 °C (Lit.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00007002
CAS-Nummer: 6203-18-5
EINECS: 228-267-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=964b8d0ff27a466c1bd1e47f2124d3da6cfee415&vid=4e69a8164fb4818d97a0b1559e5593c9ba9016ae&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (colour) Yellow to very dark yellow and orange to very dark orange
Appearance (form) Powder
Purity (HPLC area %) ≥97.5 %
Solubility (colour) Yellow to very dark yellow to very dark red
Solubility (turbidity) Clear to hazy
Solubility (method) 250 mg in 5 ml chloroform/ethanol (1:1)
Proton nmr spectrum Conforms to structure
Recommended retest period 48 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR