3,5-Dichlor-2-hydroxybenzolsulfonsäure Natriumsalz Geeignet für die Peroxidase-Analyse, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
Sodium 3,5-Dichloro-2-hydroxybenzenesulphonate, DHBS
M-Clarity™ Qualitätsstufe = MQ200
Sodium 3,5-dichloro-2-hydroxybenzenesulfonate is used in conjunction with 4-aminoantipyrine (4-AAP) and hydrogen peroxide (H2O2) for chromogenic quantitation of uric acid in serum and urine samples.
Used in conjunction with 4-aminoantipyrine (4-AAP) and hydrogen peroxide (H2O2) for chromogenic quantitation of peroxidase in coupled enzymatic reactions. Component of trinder reagent for use with peroxidase to measure generation of hydrogen peroxide in automated systems.
Formel:
C₆H₃Cl₂NaO₄S Molecular Weight: 265.05 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Room Temperature |
MDL:
MFCD00009798 CAS-Nummer: 54970-72-8 EINECS: 259-416-8 |
|
Spezifikation Testergebnisse
Appearance (Colour) | White to Off-White |
Appearance (Form) | Powder |
ICP Major Analysis Confirms Sodium Component |
Confirmed |
Purity (TLC) | ≥98 % |
Solubility (Colour) | Colorless to Faint Yellow |
Solubility (Turbidity) 5% H2O |
Clear to Slightly Hazy |
Recommended Retest Period | 2 Years |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...