3,5-Dichlor-2-hydroxybenzolsulfonsäure Natriumsalz Geeignet für die Peroxidase-Analyse, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Sodium 3,5-Dichloro-2-hydroxybenzenesulphonate, DHBS

M-Clarity™ Qualitätsstufe = MQ200

D4645-100G D4645-25G D4645-5G
D4645-100GEA 780 EUR
D4645-100G D4645-25G D4645-5G
3,5-Dichlor-2-hydroxybenzolsulfonsäure Natriumsalz Geeignet für die Peroxidase-Analyse, Sigma-Aldrich®
3,5-Dichlor-2-hydroxybenzolsulfonsäure Natriumsalz

Sodium 3,5-dichloro-2-hydroxybenzenesulfonate is used in conjunction with 4-aminoantipyrine (4-AAP) and hydrogen peroxide (H2O2) for chromogenic quantitation of uric acid in serum and urine samples.


Used in conjunction with 4-aminoantipyrine (4-AAP) and hydrogen peroxide (H2O2) for chromogenic quantitation of peroxidase in coupled enzymatic reactions. Component of trinder reagent for use with peroxidase to measure generation of hydrogen peroxide in automated systems.

Formel: C₆H₃Cl₂NaO₄S
Molecular Weight: 265.05 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Room Temperature
MDL: MFCD00009798
CAS-Nummer: 54970-72-8
EINECS: 259-416-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ed0c5d2c3e382474bc4c50d6cf825a260b093c4&vid=8ed0c25ded573abccf8cb672dee47285221f33a9&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Off-White
Appearance (Form) Powder
ICP Major Analysis
Confirms Sodium Component
Confirmed
Purity (TLC) ≥98 %
Solubility (Colour) Colorless to Faint Yellow
Solubility (Turbidity)
5% H2O
Clear to Slightly Hazy
Recommended Retest Period 2 Years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR