N,N-Diethyl-p-phenylenediamine oxalate salt, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

D5143-25G
D5143-25GEA 533 EUR
D5143-25G
N,N-Diethyl-p-phenylenediamine oxalate salt, Sigma-Aldrich®
N,N-Diethyl-p-phenylenediamine oxalate salt

N,N-Diethyl-p-phenylenediamine oxalate salt, (4-Amino-N,N-diethylaniline oxalate salt)

Formel: CH₁₆N₂
Schmelzpunkt: 145 °C (Zersetzung)
Lagertemperatur: Kühlschrank
MDL: MFCD00042025
CAS-Nummer: 142439-89-2
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54cf64212f5a327faa0c394174ed54bedfe84a3d&vid=a8b0ec3342da4607a55611f0497f2ce831e22c26&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR