6-(2,2-Dicyanovinyl)-N-(2-hydroxyethyl)-1,2,3,4-tetrahydroquinoline, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
D5928-5MG
D5928-5MGEA
323
EUR
D5928-5MG
6-(2,2-Dicyanovinyl)-N-(2-hydroxyethyl)-1,2,3,4-tetrahydroquinoline, Sigma-Aldrich®
6-(2,2-Dicyanovinyl)-N-(2-hydroxyethyl)-1,2,3,4-tetrahydroquinoline
Fluorescent molecular rotor
Formel:
C₁₅H₁₅N₃O Molecular Weight: 253,3 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00274418 CAS-Nummer: 142978-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...