Dihydroethidium ≥95%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
3,8-Diamino-5,6-dihydro-5-ethyl-6-phenylphenanthridine, Hydroethidine, DHE, 5-Ethyl-6-phenyl-5,6-dihydrophenanthridine-3,8-diamine, 2,7-Diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine
M-Clarity™ Qualitätsstufe = MQ100
D7008-10MG
D7008-10MGEA
269
EUR
D7008-10MG
Dihydroethidium ≥95%, Sigma-Aldrich®
Dihydroethidium
Redox indicator. Blue fluorescence until oxidized to ethidium. Dihydroethidium has been used as a superoxide probe in bovine pulmonary artery endothelial cell (BPAEC).
Formel:
C₂₁H₂₁N₃ Molecular Weight: 315,412 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00077335 CAS-Nummer: 104821-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...