Di-4-ANEPPS, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
D8064-5MG
D8064-5MGEA
332
EUR
D8064-5MG
Di-4-ANEPPS, Sigma-Aldrich®
Di-4-ANEPPS
4-(2-(6-(Dibutylamino)-2-naphthalenyl)ethenyl)-1-(3-sulfopropyl)pyridinium hydroxide inner salt has been used as a voltage sensitive dye in cardiac cells.
Formel:
C₂₈H₃₆N₂O₃S Lagertemperatur: Kühlschrank |
MDL:
MFCD03453609 CAS-Nummer: 90134-00-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...