3,3'-Diaminobenzidin Tetrahydrochlorid Hydrat, ISOPAC®, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: DAB HCl hydrate, 3,3',4,4'-Biphenyltetramine tetrahydrochloride hydrate, DAB tetrahydrochloride hydrate

M-Clarity™ Qualitätsstufe = MQ100

D9015-100MG
D9015-100MGEA 159 EUR
D9015-100MG
3,3'-Diaminobenzidin Tetrahydrochlorid Hydrat, ISOPAC®, Sigma-Aldrich®
3,3'-Diaminobenzidin Tetrahydrochlorid Hydrat

3,3′-Diaminobenzidine tetrahydrochloride hydrate has been used in immunohistological staining procedure and in situ hybridisation. DAB (3,3′-Diaminobenzidine) is utilised in many applications for the visualisation of peroxidase activity. In the peroxidase reaction, DAB serves as a hydrogen donor in the presence of peroxide. The oxidised DAB forms an insoluble brown end-product for use in immunohistological and immunoblotting staining procedures.


Verpackung: Weight approximate. Packaged in a 100 ml serum bottle with silicon/PTFE septum and aluminum tear seal.


Bitte beachten Injecting a compatible solvent permits preparation of any desired strength solution without exposure.

Formel: C₁₂H₁₄N₄·4HCl·H₂O
Molecular Weight: 360,11 g/mol (anhy)
Schmelzpunkt: 280 °C
Lagertemperatur: Room Temperature
MDL: MFCD08273058
CAS-Nummer: 868272-85-9
EINECS: 231-018-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8339148bf0af23ca9bd5f6f9f3d1873f4f915d0f&vid=48db9be9875801b29a7ee7b560251ccbf33c161c&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour); Off-White to Faint Red to Beige Conforms to Requirements
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Purity (TLC) ≥96%
Solubility (Turbidity), c = 50 mg/ml; Water Clear to Slightly Hazy
Solubility (Colour); Faint Pink to Amber Conforms to Requirements
Water (by Karl Fischer) 4.0 - 10.0%
Recommended Retest Period 2 Years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR