Diatrizoic acid, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 3,5-Di(acetamido)-2,4,6-triiodobenzoic Acid, 3,5-Bis(acetamido)-2,4,6-triiodobenzoic Acid, 3,5-Diacetamido-2,4,6-triiodobenzoic acid

D9268-50G
D9268-50GEA 204 EUR
D9268-50G
Diatrizoic acid, Sigma-Aldrich®
Diatrizoic acid

Diatrizoic acid has been used as a contrast-enhancing fluid for X-ray based measurements.

Formel: C₁₁H₉I₃N₂O₄
Molecular Weight: 613.92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00069960
CAS-Nummer: 117-96-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40114ef63778c3bfcfe9be805bf35a7c30557807&vid=826b0b406e63db322a8a3531d7fe53abc061bfc0&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR