4',6-Diamidin-2-phenylindol-dilactat (DAPI) ≥98% (durch HPLC) Verwendung in der nuklearen Färbung, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 2-(4-Carbamimidoylphenyl)indole-6-carboximidamide dilactate, 2-(4-Amidinophenyl)indole-6-carboxamidine dilactate

M-Clarity™ Qualitätsstufe = MQ300

D9564-10MG
D9564-10MGEA 204 EUR
D9564-10MG
4',6-Diamidin-2-phenylindol-dilactat (DAPI) ≥98% (durch HPLC) Verwendung in der nuklearen Färbung, Sigma-Aldrich®
4',6-Diamidin-2-phenylindol-dilactat (DAPI)

A high sensitivity dye used to detect single nucleic acid molecules. DAPI has been shown to detect a single DNA molecule in solution, and to detect femtograms of DNA in single cells and chloroplasts.


DAPI, dilactate has been used for the nuclear staining in cells.

Formel: C₁₆H₁₅N₅·₂C₃H₆O₃
Molecular Weight: 457,48 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 28718-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a758f10535e2e1fce756abb16b02354eb3625657&vid=53cd75c0722c6f38abfe42d59f00c63d0597dec9&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR