Mailing boxes, mini
Lieferant: DAKLAPACK
20121
DAKL20121EA 95.3 EUR
DAKL20121
Mailing boxes, mini
Versandtaschen und Transportbehälter
Mini mailing boxes are designed for sending diagnostic samples of Category B.
  • Cardboard material printed with UN-number and logo as per regulations
Standards: Approved (BAM and postal organisations) as external packaging for diagnostisc samples of Category B, as long as used in accordance with packaging instruction P650 in combination with safetybag and absorbent tube holders.