Safety bag made from PE. Ideal for sending biological samples (category B).
- Delivered pre-assembled with the top open ready for use with containers or tubes
- Easy to use: Peel and seal strip ensures a trouble free and quick fluid tight seal
- Suitable for one time use
- Easy to open by the recipient by means of an easy open tear notch
The rigid bag is compiled from 3 main components, the fluid tight bag containing a rigid blister inside an absorbing pouch capable of absorbing the contents of a 25 ml tube should any leakage occur.
Standards: Complies fully with EU packaging laws and is approved by the BVI (Belgium Packaging Institute).