Mini mailbox P650 forwarding set white
Lieferant: DAKLAPACK
520300
DAKL520300EA 664 EUR
DAKL520300
Mini mailbox P650 forwarding set white
Versandtaschen und Transportbehälter
The patented MiniMailBox system is approved for use in every country as a recognised safe way of transporting biological substances category B, UN3373 / P650.
  • Flap closure
  • Made from white cardboard
Commonly used system used by companies, hospitals and testing facilities.
Lieferumfang: The kit contains the MiniMailBox, Safetybag with bubble liner and absorbing 4 position tube holder.