Sample Bag, A5/C5, COEX
Lieferant: DAKLAPACK
9057
DAKL9057EA 39.8 EUR
DAKL9057
Sample Bag, A5/C5, COEX
Beutel Probenbeutel
Coex bag A5/C5 165×245 with a 50 mm flap is ideal for protecting various products such as clothing, small equipment and miscellaneous items during the shipment.
  • Super strong, 70 micron thick, extra thick quality
  • Equipped with a permanent closing strip
  • The coex bag is made of opaque light gray plastic with a black inside
  • The coex bag can also be used as a shipping envelope, shipping by letterbox post is possible (note the maximum thickness)