CRĒDO CUBE™ 4 Thermal Protection Containers
Lieferant: DAKLAPACK
CREDO_56L
DAKLCREDO_56LEA 3380 EUR
DAKLCREDO_56L
CRĒDO CUBE™ 4 Thermal Protection Containers
Transportbehälter Transportbehälter
The Crēdo Cube™ is a passive and reusable shipper qualified to hold chilled medical materials at a safe temperature for up to five days.
  • Superior thermal protection in a convenient mid-size container
  • Highly durable, lightweight
  • Provide an optimum volume ratio of interior payload space for products
  • Allows to ship more product in fewer containers, reducing distribution costs
This system is ideal for reducing shipping costs or for circumstances in which unexpected delays may occur.
Modular TIC™ (Thermal Isolation Chamber) walls with integrated PCM (Phase-Change Material) and VIPs (Vacuum Insulated Panels) makes conditioning in a freezer for year-round shipping easy and allows effortless packing and storage. TIC™ walls contain PCM and completely enclose the temperature-sensitive payload ensuring consistently stable performance.
Lieferumfang: The Crēdo Cube™ is a passive and reusable shipper qualified to hold chilled medical materials at a safe temperature for up to five days.