Safety spectacles, Horizon Flip Up
Lieferant: HONEYWELL SAFETY
3024042-STD
DALL3024042-STDEA 42.5 EUR
DALL3024042-STD
Safety spectacles, Horizon Flip Up
Schutzbrillen
The flip up dual lens safety glasses feature a clear impact resistant PC lens and a flip up shaded lens (IR 5).
  • Ajustable temple wires and flip-up window
  • Includes an integrated and superior sub-orbital protection and moulded sideshields
  • Abrasive resistant
  • Shade IR 5
Ideal for welders and industrial workers who require a shaded lens while working with high intensity light sources and a traditional clear impact-resistant polycarbonate lens when their task is complete.
Standards: ANSI Z87.1-2003