Cover spectacles, Horizon
Lieferant: HONEYWELL SAFETY
3024130-STD
DALL3024130-STDEA 41.1 EUR
DALL3024130-STD
Cover spectacles, Horizon
Schutzbrillen
The highly technical cover glasses (OTG) are made from a polyamide frame and moulded sideshields.
  • Easily adjustable temples
  • Integrated superior and sub-orbital protection
  • Can be equipped with four types of flip-up windows depending on the application
Frame designed to accept prescription lenses.
Weight: 34 g.