Earmuffs, QM24+®
Lieferant: HONEYWELL SAFETY
3302152
DALL3302152EA 25.8 EUR
DALL3302152
Earmuffs, QM24+®
Kapselgehörschützer Lärmschutz-Kapselgehörschützer
The dielectric QM24+® is an ultra-lightweight, three-position earmuff designed for extended wear at a valued price. Ideal for use in work environments with electricity.
  • Multiple-position headband allows wearer to select position over the head, behind the head or under the chin
  • Soft foam cushions are easy to clean with soap and water, and can be replaced for extended use
  • Alternative to cap-mounted earmuffs – wear with hard hats, face shields, respirators and other PPE
  • Dielectric construction suitable for electrical environments
SNR: 26 dB
Weight: 300 g