Analysentrichter
Lieferant: DELTALAB
19150
DELT19150EA 236 EUR
DELT19150
Analysentrichter
Trichter Filtertrichter Analysentrichter
PP, translucent.
  • With long stem and grooves
  • Rigidified by a thickened edge
  • Rapid flow due to a steep 60° angle
Designed for use with standard filter paper.