Bed pan
Lieferant: DELTALAB
19586
DELT19586EA 317 EUR
DELT19586
Bed pan
Klinisches Diagnosezubehör Klinische Probenentnahme und Probenvorbereitung Sammlung und Vorbereitung von Urinproben
Bed pan made of white PP, designed for disabled people, for the collection of urine and faeces.
  • Features rounded edges for more comfort.
  • L-shaped handle for a better handling
  • Allows a secure one-handed grip and avoids accidental spillage
  • Can be washed in automatic machines
  • Useful for hanging on handles and beds
Lieferumfang: Supplied as individually wrapped and with instructions for use.