Safety Containers
Lieferant: DELTALAB
240011
DELT240011EA 264 EUR
DELT240011
Safety Containers
Transportbehälter Flaschentragebehälter
Safety containers, made of HDPE with lid, made of PP.
  • Suitable for solid waste type II and III, enable incineration
  • Container body consists hazard warning label
It consists two lids, an entire opening, taking advantage of the total diameter of the mouth of the container, allows to throw away bigger volume waste. A partial opening used for handling syringes, contact plates, and midsize waste
Standards: In accordance with european standards ONU ADR, ISO 6706, BS 5404.