Liquid Amies, Sterile, Transporter Swab
Lieferant: DELTALAB
300284.SE
DELT300284.SEEA 1110 EUR
DELT300284.SE
Liquid Amies, Sterile, Transporter Swab
Tupfer Transport-Tupfer
This is a variation of the traditional Amies media swab where the media is presented on liquid form without agar.
  • Peel-pack dimensions: 38×210 mm
Media is supplied in its liquid form, in a sponge, suitable for direct extensions on slides.
Swabs are sterilised by radiation.
Suitable for conservation of pathogens such as:Haemophilus sp., Corynebacterium sp., Streptococcus pyogenes, Streptococcus pneumoniae, Shigella flexneri, Salmonella typhi, Brucella abortus, Staphylococcus epidermidis, Escherichia coli.
Standards: CE MDD Class IIa. CE IVD.
Bitte beachten Expiry date: 30 months after sterilisation date.