Reaktionsgefäße
Lieferant: DELTALAB
4092.1N
DELT4092.1NEA 1100 EUR
DELT4092.1N
Reaktionsgefäße
Röhrchen Reaktionsgefäße
Made of ultra clear polypropylene.
  • Autoclavable
  • Tubes have a frosted writing surface on side and on closure for better sample identification
  • Lids are easily pierceable
  • Tubes have moulded-in graduations in 100 µl increments from 0,1 to 0,6 ml
  • Withstand centrifugation up to 17000×g
  • Can be used from –80 to 121 °C
  • RNAse, DNAse and PCR inhibitors free
Caps are easy to handle and can be opened and closed with one hand.