Urine container, with boric acid
Lieferant: DELTALAB
409202.B
DELT409202.BEA 377 EUR
DELT409202.B
Urine container, with boric acid
Klinisches Diagnosezubehör Klinische Probenentnahme und Probenvorbereitung Sammlung und Vorbereitung von Urinproben
This container includes a boric acid preservative which allows the storage and transport of the urine sample to the laboratory.
  • The bacteria remain viable for 24 to 48 hours or more, depending on the type of microorganisms
  • Sterile container, sterilised by ethylene oxide
  • Boric acid quantity of 0,4 g
The boric acid supports the viability of the bacteria while preventing further multiplication. This preservative does not alter the characteristics of the urine and will not affect sample determination.
Bitte beachten *Produkte auf dieser Seite sindMore Product Information