Containers, square, with screw cap
Lieferant: DELTALAB
444608 444609 444610 444611 444612 444613 444614
DELT444608EA 782 EUR
DELT444608 DELT444609 DELT444610 DELT444611 DELT444612 DELT444613 DELT444614
Containers, square, with screw cap
Probenbehälter Universalbehälter
Containers and insert plugs made of HDPE, caps are made of PP.
  • Good chemical and temperature resistance
  • Tagging points on neck and cap for a tamper proof closure or for labelling
  • All models have moulded-in graduations
Translucent square containers with star-shaped caps.