Digital timer
Lieferant: DELTALAB
900600
DELT900600EA 130 EUR
DELT900600
Digital timer
Timer Timer, digital
Counts up and down.
  • Capacity: 24 hours
  • Alarm (60 seconds)
  • Incl. clip and small bench stand
Lieferumfang: Supplied with LR44 battery.