Microtube racks, reversible, with removable handles
Lieferant: DELTALAB
M-563A
DELTM-563AEA 202 EUR
DELTM-563A
Microtube racks, reversible, with removable handles
Gestelle Reagenzglasgestelle
PP, reversible.
  • Stackable
  • By removing the handles, units can be firmly anchored laterally to one another
The racks have two sides for different tubes: one side holds up to 80 tubes of 12 mm diameter (for example 10×75 mm or 12×75 mm tubes, microtubes, etc.) and the other side holds up to 60 microtubes, PCR, etc.