Isofreeze racks with temperature indicator
Lieferant: DELTALAB
M-572
DELTM-572EA 459 EUR
DELTM-572
Isofreeze racks with temperature indicator
Gestelle Kühlgestelle
PP, with clear lid.
  • Filled with a non toxic gel that maintains 4 °C temperature for almost four hours when the rack is removed from the freezer
  • A colour indicator begins to change from green to yellow as the temperature rises above 0 degrees