Test tube racks
Lieferant: DELTALAB
U-15128A
DELTU-15128AEA 52 EUR
DELTU-15128A
Test tube racks
Gestelle Reagenzglasgestelle
Test tube racks, made of PP, with transparent lid.
  • Excellent chemical resistance
  • Resist –196 °C and autoclavable up to 121 °C
  • Eays to clean
Available in fluorescent colours with transparent lid to help identification. Sections will hold 25 tubes up to 17 mm diameter and 125 mm height.