N-Octyl-β-D-glucopyranosid
Lieferant: G Bioscienes
|
Achtung
|
DG015
DG015EA
293
EUR
DG015
N-Octyl-β-D-glucopyranosid
N-Octyl-β-D-glucopyranosid
Formel:
C₁₄H₂₈O₆ Schmelzpunkt: 110 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00063288 CAS-Nummer: 29836-26-8 EINECS: 249-887-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...