Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamid)

Lieferant: G Bioscienes
Gefahr

Synonyme: N,N-Bis(3-D-gluconamidopropyl)cholamide

DG023
DG023EA 430 EUR
DG023
Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamid)
Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamid)
Formel: C₄₂H₇₅N₃O₁₆
Lagertemperatur: Kühlschrank
MDL: MFCD00082541
CAS-Nummer: 86303-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR