Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamid)
Lieferant: G Bioscienes
|
Gefahr
|
DG024
DG024EA
1690
EUR
DG024
Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamid)
Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamid)
Formel:
C₄₂H₇₅N₃O₁₆ Lagertemperatur: Kühlschrank |
MDL:
MFCD00082541 CAS-Nummer: 86303-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...