Desoxy-Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]desoxycholamid)

Lieferant: G Bioscienes
Achtung

Synonyme: Deoxy-bigchap

DG026
DG026EA 3560 EUR
DG026
Desoxy-Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]desoxycholamid)
Desoxy-Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]desoxycholamid)
Formel: C₄₂H₇₅N₃O₁₅
Molecular Weight: 862.05 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00161482
CAS-Nummer: 86303-23-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR