3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS)
Lieferant: G Bioscienes
Synonyme:
CHAPS
DG051
DG051EA
1550
EUR
DG051
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS)
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS)
Formel:
C₃₂H₅₈N₂O₇S Molecular Weight: 614.89 g/mol Schmelzpunkt: 116 Lagertemperatur: Kühlschrank |
MDL:
MFCD00081083 CAS-Nummer: 75621-03-3 Merck Index: 13,02052 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...