3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propansulfonat

Lieferant: G Bioscienes
Achtung

Synonyme: 3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxypropansulfonat, CHAPSO

DG053
DG053EA 1670 EUR
DG053
3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propansulfonat
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
Formel: C₃₂H₅₈N₂O₈S
Molecular Weight: 630.89 g/mol
Schmelzpunkt: 184…186 °C
Lagertemperatur: Kühlschrank
CAS-Nummer: 82473-24-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR